Anti-MESP2 Rabbit Recombinant Polyclonal Antibody
New Product
Type:
Antigen: MESP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77821-772
- 77821-771
- Antigen Symbol:
- MESP2
- MESP2
- Antigen Name:
- Mesoderm posterior protein 2 protein (218-397AA)
- Mesoderm posterior protein 2 protein (218-397AA)
- Antigen Synonyms:
- Basic helix loop helix protein MESP 2|Basic helix loop helix protein MESP2|BHLH protein MesP2|bHLHc6|Class C basic helix-loop-helix protein 6|Hypothetical class II basic helix loop helix protein MESP 2|Hypothetical class II basic helix loop helix protein MESP2|Mesoderm posterior 2|mesoderm posterior 2 homolog (mouse)|Mesoderm posterior 2 homolog|Mesoderm posterior protein 2|Mesp 2|MESP2|MESP2_HUMAN|SCDO 2|SCDO2
- Basic helix loop helix protein MESP 2|Basic helix loop helix protein MESP2|BHLH protein MesP2|bHLHc6|Class C basic helix-loop-helix protein 6|Hypothetical class II basic helix loop helix protein MESP 2|Hypothetical class II basic helix loop helix protein MESP2|Mesoderm posterior 2|mesoderm posterior 2 homolog (mouse)|Mesoderm posterior 2 homolog|Mesoderm posterior protein 2|Mesp 2|MESP2|MESP2_HUMAN|SCDO 2|SCDO2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q0VG99
- Q0VG99
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together