Anti-Toxin zeta Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Toxin zeta
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Streptococcus agalactiae
Specifications
- Catalog No:
- 77817-677
- 77817-678
- Antigen Symbol:
- Toxin zeta
- Toxin zeta
- Antigen Name:
- Recombinant Streptococcus agalactiae Toxin zeta protein (1-287AA)
- Recombinant Streptococcus agalactiae Toxin zeta protein (1-287AA)
- Antigen Synonyms:
- antibody|UDP-N-acetylglucosamine kinase|UNAG kinase|EC 2.7.1.176
- antibody|UDP-N-acetylglucosamine kinase|UNAG kinase|EC 2.7.1.176
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Streptococcus agalactiae
- Streptococcus agalactiae
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P0A4M2
- P0A4M2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together