Anti-ADAM15 Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ADAM15
Clonality: Recombinant
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77796-327
- 77796-326
- Antigen Symbol:
- ADAM15
- ADAM15
- Antigen Name:
- Disintegrin and metalloproteinase domain-containing protein 15 protein (207-452AA)
- Disintegrin and metalloproteinase domain-containing protein 15 protein (207-452AA)
- Antigen Synonyms:
- A disintegrin and metalloproteinase domain 15 (metargidin)|A disintegrin and metalloproteinase domain 15|ADA15_HUMAN|ADAM 15|ADAM metallopeptidase domain 15|Adam15|and cysteine-rich protein 15|Disintegrin and metalloproteinase domain-containing protein 15|disintegrin-like|EC 3.4.24.|MDC 15|MDC-15|MDC15|Metalloprotease RGD disintegrin protein|Metalloproteinase like disintegrin like and cysteine rich protein 15|Metalloproteinase-like|Metargidin
- A disintegrin and metalloproteinase domain 15 (metargidin)|A disintegrin and metalloproteinase domain 15|ADA15_HUMAN|ADAM 15|ADAM metallopeptidase domain 15|Adam15|and cysteine-rich protein 15|Disintegrin and metalloproteinase domain-containing protein 15|disintegrin-like|EC 3.4.24.|MDC 15|MDC-15|MDC15|Metalloprotease RGD disintegrin protein|Metalloproteinase like disintegrin like and cysteine rich protein 15|Metalloproteinase-like|Metargidin
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13444
- Q13444
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together