Anti-Thbs3 Rabbit Recombinant Antibody
Catalog # 77802-490
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Recombinant Mouse Thrombospondin-3 protein (24-350AA)
- Antigen Symbol:Thbs3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse,Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q05895
- Antigen Synonyms:Thbs3|Tsp3|Thrombospondin-3
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: Thbs3
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Human