Anti-SDF4 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: SDF4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77835-194
- 77835-193
- Antigen Symbol:
- SDF4
- SDF4
- Antigen Name:
- 45 kDa calcium-binding protein (37-362AA)
- 45 kDa calcium-binding protein (37-362AA)
- Antigen Synonyms:
- 45 kDa calcium-binding protein|Cab45|CAB45_HUMAN|Calcium binding protein|PSEC0034|SDF 4|SDF-4|SDF4|Stromal cell derived factor 4|Stromal cell-derived factor 4
- 45 kDa calcium-binding protein|Cab45|CAB45_HUMAN|Calcium binding protein|PSEC0034|SDF 4|SDF-4|SDF4|Stromal cell derived factor 4|Stromal cell-derived factor 4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BRK5
- Q9BRK5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together