Anti-POLR2I Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: POLR2I
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77794-772
- 77794-771
- Antigen Symbol:
- POLR2I
- POLR2I
- Antigen Name:
- DNA-directed RNA polymerase II subunit RPB9 protein (1-125AA)
- DNA-directed RNA polymerase II subunit RPB9 protein (1-125AA)
- Antigen Synonyms:
- DNA directed RNA polymerase II 14.5 kDa polypeptide|DNA directed RNA polymerase II polypeptide I|DNA directed RNA polymerase II subunit I|DNA directed RNA polymerase II subunit RPB9|DNA-directed RNA polymerase II subunit I|DNA-directed RNA polymerase II subunit RPB9|hRPB14.5|POLR2I|Polymerase (RNA) II (DNA directed) polypeptide I 14.5kDa|RNA polymerase II 14.5 kDa subunit|RNA polymerase II subunit B9|RPB14.5|RPB9|RPB9_HUMAN
- DNA directed RNA polymerase II 14.5 kDa polypeptide|DNA directed RNA polymerase II polypeptide I|DNA directed RNA polymerase II subunit I|DNA directed RNA polymerase II subunit RPB9|DNA-directed RNA polymerase II subunit I|DNA-directed RNA polymerase II subunit RPB9|hRPB14.5|POLR2I|Polymerase (RNA) II (DNA directed) polypeptide I 14.5kDa|RNA polymerase II 14.5 kDa subunit|RNA polymerase II subunit B9|RPB14.5|RPB9|RPB9_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P36954
- P36954
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together