Anti-NEK10 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NEK10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-180
- 77830-179
- Antigen Symbol:
- NEK10
- NEK10
- Antigen Name:
- Serine/threonine-protein kinase Nek10 protein (1-104AA)
- Serine/threonine-protein kinase Nek10 protein (1-104AA)
- Antigen Synonyms:
- NEK10|Serine/threonine-protein kinase Nek10|EC 2.7.11.1|Never in mitosis A-related kinase 10|NimA-related protein kinase 10
- NEK10|Serine/threonine-protein kinase Nek10|EC 2.7.11.1|Never in mitosis A-related kinase 10|NimA-related protein kinase 10
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6ZWH5
- Q6ZWH5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together