Anti-PCCA Rabbit Recombinant Polyclonal Antibody
Catalog # 77793-343
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Propionyl-CoA carboxylase alpha chain, mitochondrial protein (585-720AA)
- Antigen Symbol:PCCA
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P05165
- Antigen Synonyms:mitochondrial|Pcca|pccA COMPLEMENTATION GROUP|PCCA_HUMAN|PCCase subunit alpha|Propanoyl CoA:carbon dioxide ligase subunit alpha|Propanoyl-CoA:carbon dioxide ligase subunit alpha|Propionyl CoA carboxylase alpha chain|Propionyl CoA carboxylase alpha chain mitochondrial|Propionyl CoA carboxylase alpha polypeptide|Propionyl coenzyme A carboxylase alpha polypeptide|Propionyl-CoA carboxylase alpha chain
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended Dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: PCCA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse