Anti-ARPC2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ARPC2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77806-686
- 77806-685
- Antigen Symbol:
- ARPC2
- ARPC2
- Antigen Name:
- Actin-related protein 2/3 complex subunit 2 protein (1-250AA)
- Actin-related protein 2/3 complex subunit 2 protein (1-250AA)
- Antigen Synonyms:
- Actin related protein 2/3 complex subunit 2 34kDa|Actin related protein 2/3 complex subunit 2|Actin-related protein 2/3 complex subunit 2|ARC 34|ARC34|Arp2/3 complex 34 kDa subunit|ARP2/3 protein compex subunit 34|ARPC 2|Arpc2|ARPC2_HUMAN|p34 Arc|p34-ARC|p34Arc|PNAS 139|PNAS139|PRO 2446|PRO2446
- Actin related protein 2/3 complex subunit 2 34kDa|Actin related protein 2/3 complex subunit 2|Actin-related protein 2/3 complex subunit 2|ARC 34|ARC34|Arp2/3 complex 34 kDa subunit|ARP2/3 protein compex subunit 34|ARPC 2|Arpc2|ARPC2_HUMAN|p34 Arc|p34-ARC|p34Arc|PNAS 139|PNAS139|PRO 2446|PRO2446
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15144
- O15144
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together