Anti-YIF1B Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: YIF1B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77827-004
- 77827-003
- Antigen Symbol:
- YIF1B
- YIF1B
- Antigen Name:
- Protein YIF1B protein (1-153AA)
- Protein YIF1B protein (1-153AA)
- Antigen Synonyms:
- FinGER8| FLJ50074| Protein YIF1B| YIF1B| YIF1B_HUMAN| Yip1 interacting factor homolog B (S. cerevisiae)| Yip1 interacting factor homolog B| YIP1-interacting factor homolog B| Yip1b
- FinGER8| FLJ50074| Protein YIF1B| YIF1B| YIF1B_HUMAN| Yip1 interacting factor homolog B (S. cerevisiae)| Yip1 interacting factor homolog B| YIP1-interacting factor homolog B| Yip1b
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5BJH7
- Q5BJH7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together