Anti-TET2 Rabbit Polyclonal Antibody
Catalog # 77802-244
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:TET2_HUMAN
- Antigen Symbol:TET2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q6N021
- Antigen Synonyms:FLJ20032|KIAA1546|MDS|Methylcytosine dioxygenase TET2|Nbla00191|Probable methylcytosine dioxygenase TET2|Protein Ayu17 449|Tet 2|Tet methylcytosine dioxygenase 2|Tet oncogene 2|Tet oncogene family member 2|TET2|TET2_HUMAN
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TET2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human