Anti-MID1IP1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MID1IP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-609
- 77838-608
- Antigen Symbol:
- MID1IP1
- MID1IP1
- Antigen Name:
- Mid1-interacting protein 1 protein (1-183AA)
- Mid1-interacting protein 1 protein (1-183AA)
- Antigen Synonyms:
- 3110038L01Rik|FLJ10386|G12 like|Gastrulation specific G12 like protein|Gastrulation-specific G12-like protein|M1IP1_HUMAN|MGC72582|MID1 interacting G12 like protein|MID1 interacting protein 1 (gastrulation specific G12 like)|Mid1-interacting G12-like protein|Mid1-interacting protein 1|Mid1ip1|MIG12|OTTMUSP00000018143|OTTMUSP00000018222|OTTMUSP00000018284|OTTMUSP00000018285|Protein STRAIT11499|Protein STRAIT11499 homolog|RP23-130J1.1|S14R|Slap|Spot 14 like androgen inducible protein|Spot 14 related protein|Spot 14-R|Spot 14-related protein|STRAIT11499|THRSPL
- 3110038L01Rik|FLJ10386|G12 like|Gastrulation specific G12 like protein|Gastrulation-specific G12-like protein|M1IP1_HUMAN|MGC72582|MID1 interacting G12 like protein|MID1 interacting protein 1 (gastrulation specific G12 like)|Mid1-interacting G12-like protein|Mid1-interacting protein 1|Mid1ip1|MIG12|OTTMUSP00000018143|OTTMUSP00000018222|OTTMUSP00000018284|OTTMUSP00000018285|Protein STRAIT11499|Protein STRAIT11499 homolog|RP23-130J1.1|S14R|Slap|Spot 14 like androgen inducible protein|Spot 14 related protein|Spot 14-R|Spot 14-related protein|STRAIT11499|THRSPL
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NPA3
- Q9NPA3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together