Anti-SLC5A11 Rabbit Polyclonal Antibody
Catalog # 77800-121
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Solute carrier family 5 member 11
- Antigen Symbol:SLC5A11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q8WWX8
- Antigen Synonyms:homolog of rabbit KST1|KST1|Na(+)/myo-inositol cotransporter 2|na+/myo-inositol cotransporter 2|putative sodium-coupled cotransporter RKST1|RKST1|SC5AB_HUMAN|SGLT6|SLC5A11|SMIT2|Sodium-dependent glucose cotransporter|Sodium/glucose cotransporter KST1|Sodium/myo-inositol cotransporter 2|Sodium/myo-inositol transporter 2|solute carrier family 5 (sodium/glucose cotransporter)|member 11|Solute carrier family 5 member 11
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: SLC5A11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat