To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antigen Name:Synthesized peptide derived from MEK-4 around the non-phosphorylation site of S257.
- Antigen Symbol:MAP2K4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Form:Liquid
- Gene ID:P45985
- Antigen Synonyms:c Jun N terminal kinase kinase 1|C-JUN N-terminal kinase kinase 1|Dual specificity mitogen activated protein kinase kinase 4|Dual specificity mitogen-activated protein kinase kinase 4|JNK Activated Kinase 1|JNK activating kinase 1|JNK-activating kinase 1|JNKK|JNKK1|MAP kinase kinase 4|Map2k4|MAPK ERK kinase 4|MAPK/ERK kinase 4|MAPKK 4|MAPKK4|MEK 4|MEK4|Mitogen activated protein kinase kinase 4|MKK 4|MKK4|MP2K4_HUMAN|PRKMK4|SAPK ERK kinase 1|SAPK/ERK kinase 1|SAPKK 1|SAPKK1|SEK1|SERK1|SKK1|Stress activated protein kinase kinase 1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC: 1:100 to 1:300, IF: 1:200 to 1:1000, ELISA:1:10000
Type:
Antigen: MAP2K4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat