Anti-CXCL11 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CXCL11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77809-481
- 77809-480
- Antigen Symbol:
- CXCL11
- CXCL11
- Antigen Name:
- C-X-C motif chemokine 11 protein (22-94AA)
- C-X-C motif chemokine 11 protein (22-94AA)
- Antigen Synonyms:
- b R1|b-R1|Beta-R1|betaR1|C-X-C motif chemokine 11|Chemokine (C-X-C motif) ligand 11|Chemokine C-X-C motif ligand 11|CXC11|CXCL11|CXL11_HUMAN|H174|I TAC|I-TAC|Interferon gamma inducible protein 9|Interferon gamma-inducible protein 9|Interferon inducible T cell alpha chemoattractant|Interferon-inducible T-cell a chemoattractant I-TAC|Interferon-inducible T-cell alpha chemoattractant|IP 9|IP-9|IP9|ITAC|MGC102770|SCYB11|SCYB9B|Small inducible cytokine B11|Small inducible cytokine subfamily B (Cys-X-Cys) member 11|Small inducible cytokine subfamily B (Cys-X-Cys) member 9B|Small inducible cytokine subfamily B|member 11|member 9B|Small-inducible cytokine B11
- b R1|b-R1|Beta-R1|betaR1|C-X-C motif chemokine 11|Chemokine (C-X-C motif) ligand 11|Chemokine C-X-C motif ligand 11|CXC11|CXCL11|CXL11_HUMAN|H174|I TAC|I-TAC|Interferon gamma inducible protein 9|Interferon gamma-inducible protein 9|Interferon inducible T cell alpha chemoattractant|Interferon-inducible T-cell a chemoattractant I-TAC|Interferon-inducible T-cell alpha chemoattractant|IP 9|IP-9|IP9|ITAC|MGC102770|SCYB11|SCYB9B|Small inducible cytokine B11|Small inducible cytokine subfamily B (Cys-X-Cys) member 11|Small inducible cytokine subfamily B (Cys-X-Cys) member 9B|Small inducible cytokine subfamily B|member 11|member 9B|Small-inducible cytokine B11
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14625
- O14625
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together