Anti-RAMP2 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: RAMP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77796-613
- 77796-612
- Antigen Symbol:
- RAMP2
- RAMP2
- Antigen Name:
- Receptor activity-modifying protein 2 protein (43-145AA)
- Receptor activity-modifying protein 2 protein (43-145AA)
- Antigen Synonyms:
- Calcitonin receptor like receptor activity modifying protein 2|Calcitonin-receptor-like receptor activity-modifying protein 2|CRLR activity modifying protein 2|CRLR activity-modifying protein 2|OTTMUSP00000002615|OTTMUSP00000037624|RAMP2_HUMAN|Receptor (calcitonin) activity modifying protein 2|Receptor activity modifying protein 2|Receptor activity modifying protein 2 isoform|Receptor activity-modifying protein 2|RP23-281C18.6
- Calcitonin receptor like receptor activity modifying protein 2|Calcitonin-receptor-like receptor activity-modifying protein 2|CRLR activity modifying protein 2|CRLR activity-modifying protein 2|OTTMUSP00000002615|OTTMUSP00000037624|RAMP2_HUMAN|Receptor (calcitonin) activity modifying protein 2|Receptor activity modifying protein 2|Receptor activity modifying protein 2 isoform|Receptor activity-modifying protein 2|RP23-281C18.6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60895
- O60895
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together