Anti-ZMPSTE24 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: ZMPSTE24
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77804-985
- 77804-986
- Antigen Symbol:
- ZMPSTE24
- ZMPSTE24
- Antigen Name:
- CAAX prenyl protease 1 homolog protein (217-347AA)
- CAAX prenyl protease 1 homolog protein (217-347AA)
- Antigen Synonyms:
- CAAX prenyl protease 1 homolog|FACE-1|FACE1|FACE1_HUMAN|Farnesylated proteins converting enzyme 1|Farnesylated proteins-converting enzyme 1|Prenyl protein specific endoprotease 1|Prenyl protein-specific endoprotease 1|STE24|Zinc metalloproteinase Ste24 homolog|zmpste24
- CAAX prenyl protease 1 homolog|FACE-1|FACE1|FACE1_HUMAN|Farnesylated proteins converting enzyme 1|Farnesylated proteins-converting enzyme 1|Prenyl protein specific endoprotease 1|Prenyl protein-specific endoprotease 1|STE24|Zinc metalloproteinase Ste24 homolog|zmpste24
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75844
- O75844
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together