Anti-NKIRAS2 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NKIRAS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-887
- 77838-886
- Antigen Symbol:
- NKIRAS2
- NKIRAS2
- Antigen Name:
- NF-kappa-B inhibitor-interacting Ras-like protein 2 protein (1-191AA)
- NF-kappa-B inhibitor-interacting Ras-like protein 2 protein (1-191AA)
- Antigen Synonyms:
- DKFZP434N1526|I kappa B interacting Ras like protein 2|I-kappa-B-interacting Ras-like protein 2|Kappa B Ras protein 2|Kappa B-Ras protein 2|KappaB Ras2|KappaB-Ras2|KBRAS 2|KBRAS2|KBRS2_HUMAN|MGC74742|NF kappa B inhibitor interacting Ras like protein 2|NF-kappa-B inhibitor-interacting Ras-like protein 2|NFKB inhibitor interacting Ras like 2|NFKB inhibitor interacting Ras like protein 2|NKIRAS 2|NKIRAS2
- DKFZP434N1526|I kappa B interacting Ras like protein 2|I-kappa-B-interacting Ras-like protein 2|Kappa B Ras protein 2|Kappa B-Ras protein 2|KappaB Ras2|KappaB-Ras2|KBRAS 2|KBRAS2|KBRS2_HUMAN|MGC74742|NF kappa B inhibitor interacting Ras like protein 2|NF-kappa-B inhibitor-interacting Ras-like protein 2|NFKB inhibitor interacting Ras like 2|NFKB inhibitor interacting Ras like protein 2|NKIRAS 2|NKIRAS2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NYR9
- Q9NYR9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together