Anti-TRIM71 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TRIM71
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77825-944
- 77825-945
- Antigen Symbol:
- TRIM71
- TRIM71
- Antigen Name:
- E3 ubiquitin-protein ligase TRIM71 protein (125-209AA)
- E3 ubiquitin-protein ligase TRIM71 protein (125-209AA)
- Antigen Synonyms:
- 2610206G21Rik|Abnormal cell LINeage LIN 41|AL022943|E3 ubiquitin-protein ligase TRIM71|Gm1127|Homolog of C. elegans Lin 41|LIN 41|LIN41|LIN41_HUMAN|mlin 41|Protein lin 41 homolog|Protein lin-41 homolog|RGD1566388|Ripply2|TRIM 71|TRIM71|Tripartite motif containing 71|Tripartite motif-containing protein 71
- 2610206G21Rik|Abnormal cell LINeage LIN 41|AL022943|E3 ubiquitin-protein ligase TRIM71|Gm1127|Homolog of C. elegans Lin 41|LIN 41|LIN41|LIN41_HUMAN|mlin 41|Protein lin 41 homolog|Protein lin-41 homolog|RGD1566388|Ripply2|TRIM 71|TRIM71|Tripartite motif containing 71|Tripartite motif-containing protein 71
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q2Q1W2
- Q2Q1W2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together