Anti-TOX Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TOX
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-334
- 77803-334
- Antigen Symbol:
- TOX
- TOX
- Antigen Name:
- Thymocyte selection-associated high mobility group box protein TOX protein (141-288AA)
- Thymocyte selection-associated high mobility group box protein TOX protein (141-288AA)
- Antigen Synonyms:
- KIAA0808|Thymocyte selection-associated high mobility group box|Thymocyte selection-associated high mobility group box protein TOX|Thymus high mobility group box protein TOX|Thymus high mobility group box protein, mouse, homolog of TOX1|TOX 1|Tox|TOX_HUMAN|TOX1
- KIAA0808|Thymocyte selection-associated high mobility group box|Thymocyte selection-associated high mobility group box protein TOX|Thymus high mobility group box protein TOX|Thymus high mobility group box protein, mouse, homolog of TOX1|TOX 1|Tox|TOX_HUMAN|TOX1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O94900
- O94900
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together