Anti-SMNDC1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SMNDC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77800-338
- 77800-339
- Antigen Symbol:
- SMNDC1
- SMNDC1
- Antigen Name:
- Survival of motor neuron-related-splicing factor 30 protein (1-238AA)
- Survival of motor neuron-related-splicing factor 30 protein (1-238AA)
- Antigen Synonyms:
- 30 kDa splicing factor SMNrp|MGC106917|MGC112663|SMN related protein|SMN-related protein|smndc1|SMNR|SPF30|SPF30_HUMAN|Splicing factor 30, survival of motor neuron-related|Survival motor neuron domain containing 1|Survival motor neuron domain containing protein 1|Survival motor neuron domain-containing protein 1|Survival of motor neuron related splicing factor 30|Survival of motor neuron-related-splicing factor 30
- 30 kDa splicing factor SMNrp|MGC106917|MGC112663|SMN related protein|SMN-related protein|smndc1|SMNR|SPF30|SPF30_HUMAN|Splicing factor 30, survival of motor neuron-related|Survival motor neuron domain containing 1|Survival motor neuron domain containing protein 1|Survival motor neuron domain-containing protein 1|Survival of motor neuron related splicing factor 30|Survival of motor neuron-related-splicing factor 30
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75940
- O75940
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together