Anti-CYP2U1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CYP2U1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77831-826
- 77831-825
- Antigen Symbol:
- CYP2U1
- CYP2U1
- Antigen Name:
- Cytochrome P450 2U1 protein (363-496AA)
- Cytochrome P450 2U1 protein (363-496AA)
- Antigen Synonyms:
- CP2U1_HUMAN|CYP2U 1 |CYP2U1|Cytochrome P450 2U1|Cytochrome P450|family 2|subfamily U|polypeptide 1|EC 1.14.14.1 |P450TEC
- CP2U1_HUMAN|CYP2U 1 |CYP2U1|Cytochrome P450 2U1|Cytochrome P450|family 2|subfamily U|polypeptide 1|EC 1.14.14.1 |P450TEC
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7Z449
- Q7Z449
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together