Anti-C1QTNF1 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: C1QTNF1
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77838-081
- 77838-080
- Antigen Symbol:
- C1QTNF1
- C1QTNF1
- Antigen Name:
- Complement C1q tumor necrosis factor-related protein 1 protein (26-281AA)
- Complement C1q tumor necrosis factor-related protein 1 protein (26-281AA)
- Antigen Synonyms:
- C1q and tumor necrosis factor related protein 1| C1QT1_HUMAN| C1QTNF1| Complement C1q tumor necrosis factor related protein 1 | Complement C1q tumor necrosis factor-related protein 1| CTRP 1| CTRP1| FLJ90694| G protein coupled receptor interacting protein 1| G protein-coupled receptor-interacting protein| GIP| ZSIG37
- C1q and tumor necrosis factor related protein 1| C1QT1_HUMAN| C1QTNF1| Complement C1q tumor necrosis factor related protein 1 | Complement C1q tumor necrosis factor-related protein 1| CTRP 1| CTRP1| FLJ90694| G protein coupled receptor interacting protein 1| G protein-coupled receptor-interacting protein| GIP| ZSIG37
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BXJ1
- Q9BXJ1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together