Anti-ADAM19 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: ADAM19
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-501
- 77840-502
- Antigen Symbol:
- ADAM19
- ADAM19
- Antigen Name:
- Disintegrin and metalloproteinase domain-containing protein 19 protein (203-460AA)
- Disintegrin and metalloproteinase domain-containing protein 19 protein (203-460AA)
- Antigen Synonyms:
- ADAM19|MLTNB|FKSG34Disintegrin and metalloproteinase domain-containing protein 19|ADAM 19|EC 3.4.24.-|Meltrin-beta|Metalloprotease and disintegrin dendritic antigen marker|MADDAM
- ADAM19|MLTNB|FKSG34Disintegrin and metalloproteinase domain-containing protein 19|ADAM 19|EC 3.4.24.-|Meltrin-beta|Metalloprotease and disintegrin dendritic antigen marker|MADDAM
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H013
- Q9H013
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together