Anti-CAPN10 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CAPN10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-324
- 77836-325
- Antigen Symbol:
- CAPN10
- CAPN10
- Antigen Name:
- Calpain-10 protein (1-287AA)
- Calpain-10 protein (1-287AA)
- Antigen Synonyms:
- Calcium activated neutral protease|Calcium activated neutral proteinase 10|Calcium-activated neutral proteinase 10|calpain like protease CAPN10|Calpain-10|Calpain10|CAN10_HUMAN|CANP 10|CANP10|CAPN 10|CAPN10|EC 3.4.22.|KIAA1845|NIDDM1
- Calcium activated neutral protease|Calcium activated neutral proteinase 10|Calcium-activated neutral proteinase 10|calpain like protease CAPN10|Calpain-10|Calpain10|CAN10_HUMAN|CANP 10|CANP10|CAPN 10|CAPN10|EC 3.4.22.|KIAA1845|NIDDM1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HC96
- Q9HC96
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together