Anti-SLC1A1 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000
Type:
Antigen: SLC1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77809-796
- 77809-795
- Antigen Symbol:
- SLC1A1
- SLC1A1
- Antigen Name:
- Synthetic peptide of SLC1A1
- Synthetic peptide of SLC1A1
- Antigen Synonyms:
- EAA3_HUMAN|EAAC 1|EAAC1|EAAT 3|Excitatory amino acid carrier 1 |Excitatory amino acid carrier1|Excitatory amino acid transporter 3|Excitatory amino acid transporter3|Excitatory amino-acid carrier 1|GLUTAMATE TRANSPORTER|HIGH-AFFINITY|MEAAC 1|MEAAC1|Neuronal and epithelial glutamate transporter|REAAC 1|REAAC1|Slc1 a1|Slc1a 1|SLC1A1|Sodium dependent glutamate/aspartate transporter 3|Sodium-dependent glutamate/aspartate transporter 3|Solute carrier family 1 (neuronal / epithelial high affinity glutamate transporter|system Xag)|member 1|SOLUTE CARRIER FAMILY 1 (NEURONAL/EPITHELIAL HIGH AFFINITY GLUTAMATE TRANSPORTER)|MEMBER 1|Solute carrier family 1 (neuronal/epithelial high affinity glutamate transporter|Solute carrier family 1 member 1|Solute carrier family 1
- EAA3_HUMAN|EAAC 1|EAAC1|EAAT 3|Excitatory amino acid carrier 1 |Excitatory amino acid carrier1|Excitatory amino acid transporter 3|Excitatory amino acid transporter3|Excitatory amino-acid carrier 1|GLUTAMATE TRANSPORTER|HIGH-AFFINITY|MEAAC 1|MEAAC1|Neuronal and epithelial glutamate transporter|REAAC 1|REAAC1|Slc1 a1|Slc1a 1|SLC1A1|Sodium dependent glutamate/aspartate transporter 3|Sodium-dependent glutamate/aspartate transporter 3|Solute carrier family 1 (neuronal / epithelial high affinity glutamate transporter|system Xag)|member 1|SOLUTE CARRIER FAMILY 1 (NEURONAL/EPITHELIAL HIGH AFFINITY GLUTAMATE TRANSPORTER)|MEMBER 1|Solute carrier family 1 (neuronal/epithelial high affinity glutamate transporter|Solute carrier family 1 member 1|Solute carrier family 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P43005
- P43005
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together