Anti-RXFP3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: RXFP3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-763
- 77838-762
- Antigen Symbol:
- RXFP3
- RXFP3
- Antigen Name:
- Relaxin-3 receptor 1 protein (1-80AA)
- Relaxin-3 receptor 1 protein (1-80AA)
- Antigen Synonyms:
- G protein coupled receptor GPCR135|G protein coupled receptor SALPR|G protein-coupled receptor SALPR|G-protein coupled receptor GPCR135|GPCR 135|GPCR135|Gprotein coupled receptor SALPR|Relaxin 3 receptor 1|Relaxin 3 receptor1|Relaxin 3/INSL7 receptor 1|Relaxin 3/INSL7 receptor1|Relaxin family peptide receptor 3|Relaxin family peptide receptor3|Relaxin-3 receptor 1|Relaxin/insulin like family peptide receptor 3|Relaxin3 receptor 1|Relaxin3 receptor1|RL3 R1|RL3R 1|RL3R1|RL3R1_HUMAN|RLN3 receptor 1|RLN3 receptor1|RLN3R1|RXFP3|RXFPR3|SALP R|SALPR|somatostatin and angiotensin like peptide receptor|Somatostatin- and angiotensin-like peptide receptor
- G protein coupled receptor GPCR135|G protein coupled receptor SALPR|G protein-coupled receptor SALPR|G-protein coupled receptor GPCR135|GPCR 135|GPCR135|Gprotein coupled receptor SALPR|Relaxin 3 receptor 1|Relaxin 3 receptor1|Relaxin 3/INSL7 receptor 1|Relaxin 3/INSL7 receptor1|Relaxin family peptide receptor 3|Relaxin family peptide receptor3|Relaxin-3 receptor 1|Relaxin/insulin like family peptide receptor 3|Relaxin3 receptor 1|Relaxin3 receptor1|RL3 R1|RL3R 1|RL3R1|RL3R1_HUMAN|RLN3 receptor 1|RLN3 receptor1|RLN3R1|RXFP3|RXFPR3|SALP R|SALPR|somatostatin and angiotensin like peptide receptor|Somatostatin- and angiotensin-like peptide receptor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NSD7
- Q9NSD7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together