Anti-KCNJ8 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KCNJ8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-174
- 77822-173
- Antigen Symbol:
- KCNJ8
- KCNJ8
- Antigen Name:
- ATP-sensitive inward rectifier potassium channel 8 protein (299-424AA)
- ATP-sensitive inward rectifier potassium channel 8 protein (299-424AA)
- Antigen Synonyms:
- inwardly rectifying subfamily J member 8|ATP sensitive inward rectifier potassium channel 8|ATP-sensitive inward rectifier potassium channel 8|Inward rectifier K(+) channel Kir6.1|Inwardly rectifier K(+) channel Kir6.1|Inwardly rectifying potassium channel Kir6.1|IRK 8|IRK8|Kcnj8|KCNJ8_HUMAN|kir 6.1|Potassium channel|potassium inwardly rectifying channel J8|uKATP 1|uKATP-1
- inwardly rectifying subfamily J member 8|ATP sensitive inward rectifier potassium channel 8|ATP-sensitive inward rectifier potassium channel 8|Inward rectifier K(+) channel Kir6.1|Inwardly rectifier K(+) channel Kir6.1|Inwardly rectifying potassium channel Kir6.1|IRK 8|IRK8|Kcnj8|KCNJ8_HUMAN|kir 6.1|Potassium channel|potassium inwardly rectifying channel J8|uKATP 1|uKATP-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15842
- Q15842
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together