Anti-PDE2A Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PDE2A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77793-555
- 77793-556
- Antigen Symbol:
- PDE2A
- PDE2A
- Antigen Name:
- cGMP-dependent 3\',5\'-cyclic phosphodiesterase protein (15-226AA)
- cGMP-dependent 3\',5\'-cyclic phosphodiesterase protein (15-226AA)
- Antigen Synonyms:
- 5''-cyclic phosphodiesterase|cGMP dependent 3'5' cyclic phosphodiesterase|cGMP stimulated phosphodiesterase 1|cGMP stimulated phosphodiesterase 4|cGMP-dependent 3''|cGMP-stimulated phosphodiesterase 2|CGS PDE|CGS-PDE|cGSPDE|Cyclic GMP stimulated phosphodiesterase|Cyclic GMP-stimulated phosphodiesterase|Human cGMP stimulated 3'5' cyclic nucleotide phosphodiesterase|PDE 2A|PDE 2A1|PDE 2A4|PDE2A|PDE2A_HUMAN|PDE2A1|PDE2A4|phosphodiesterase 2A|Phosphodiesterase 2A cGMP stimulated
- 5''-cyclic phosphodiesterase|cGMP dependent 3'5' cyclic phosphodiesterase|cGMP stimulated phosphodiesterase 1|cGMP stimulated phosphodiesterase 4|cGMP-dependent 3''|cGMP-stimulated phosphodiesterase 2|CGS PDE|CGS-PDE|cGSPDE|Cyclic GMP stimulated phosphodiesterase|Cyclic GMP-stimulated phosphodiesterase|Human cGMP stimulated 3'5' cyclic nucleotide phosphodiesterase|PDE 2A|PDE 2A1|PDE 2A4|PDE2A|PDE2A_HUMAN|PDE2A1|PDE2A4|phosphodiesterase 2A|Phosphodiesterase 2A cGMP stimulated
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00408
- O00408
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together