Anti-ALDOB Rabbit Polyclonal Antibody
Catalog # 77827-342
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from internal of ALDOB
- Antigen Symbol:ALDOB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P05062
- Antigen Synonyms:ALDB|ALDO B|ALDO2|ALDOB|ALDOB_HUMAN|Aldolase 2|Aldolase B|Aldolase B fructose bisphosphate|Aldolase2|AldolaseB|EC 4.1.2.13|Fructose bisphosphate aldolase B|Fructose-bisphosphate aldolase B|Liver type aldolase|Liver-type aldolase|MS1077
- Storage Buffer:PBS, 150 mM NaCl, 0.02% Sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: ALDOB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat