Anti-NELL2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NELL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-693
- 77834-692
- Antigen Symbol:
- NELL2
- NELL2
- Antigen Name:
- Protein kinase C-binding protein NELL2 protein (511-637AA)
- Protein kinase C-binding protein NELL2 protein (511-637AA)
- Antigen Synonyms:
- A330108N19Rik|Mel91|MEL91 protein|NEL like 2 (chicken)|NEL-like protein 2|Nel-related protein 2|NELL2|NELL2_HUMAN|Neural epidermal growth factor like 2|NRP2|Protein kinase C-binding protein NELL2|R75516
- A330108N19Rik|Mel91|MEL91 protein|NEL like 2 (chicken)|NEL-like protein 2|Nel-related protein 2|NELL2|NELL2_HUMAN|Neural epidermal growth factor like 2|NRP2|Protein kinase C-binding protein NELL2|R75516
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99435
- Q99435
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together