Anti-gpd-1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: gpd-1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Caenorhabditis elegans
Specifications
- Catalog No:
- 77817-470
- 77817-471
- Antigen Symbol:
- gpd-1
- gpd-1
- Antigen Name:
- Recombinant Caenorhabditis elegans Glyceraldehyde-3-phosphate dehydrogenase 1 protein (1-341AA)
- Recombinant Caenorhabditis elegans Glyceraldehyde-3-phosphate dehydrogenase 1 protein (1-341AA)
- Antigen Synonyms:
- gpd-1|T09F3.3Glyceraldehyde-3-phosphate dehydrogenase 1|GAPDH-1|EC 1.2.1.12
- gpd-1|T09F3.3Glyceraldehyde-3-phosphate dehydrogenase 1|GAPDH-1|EC 1.2.1.12
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Caenorhabditis elegans
- Caenorhabditis elegans
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P04970
- P04970
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together