Anti-rtcB Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: rtcB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Escherichia coli
Specifications
- Catalog No:
- 77816-361
- 77816-362
- Antigen Symbol:
- rtcB
- rtcB
- Antigen Name:
- Recombinant Escherichia coli RNA-splicing ligase RtcB protein (1-408AA)
- Recombinant Escherichia coli RNA-splicing ligase RtcB protein (1-408AA)
- Antigen Synonyms:
- yhgL|b3421|JW3384|RNA-splicing ligase RtcB|EC 6.5.1.8|3'-phosphate/5'-hydroxy nucleic acid ligase
- yhgL|b3421|JW3384|RNA-splicing ligase RtcB|EC 6.5.1.8|3'-phosphate/5'-hydroxy nucleic acid ligase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- E. coli
- E. coli
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P46850
- P46850
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together