Anti-NPL Rabbit Recombinant Polyclonal Antibody
77839-184
New Product
- Antigen Name:N-acetylneuraminate lyase protein (17-77AA)
- Antigen Symbol:NPL
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9BXD5
- Antigen Synonyms:0610033B02Rik|C112|C1orf13|Dihydrodipicolinate synthase|Dihydrodipicolinate synthetase homolog 1|MGC149582|MGC61869|N-acetylneuraminate lyase|N-acetylneuraminate pyruvate-lyase|N-acetylneuraminic acid aldolase|NAL|NALase|NPL|NPL_HUMAN|NPL1|RP11-249O6.1|Sialate lyase|Sialate-pyruvate lyase|Sialic acid aldolase|Sialic acid lyase
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB:1:500 to 1:5000, IF:1:50 to 1:200
Type:
Antigen: NPL
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse