Anti-UROS Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UROS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77804-075
- 77804-074
- Antigen Symbol:
- UROS
- UROS
- Antigen Name:
- Uroporphyrinogen-III synthase protein (1-265AA)
- Uroporphyrinogen-III synthase protein (1-265AA)
- Antigen Synonyms:
- congenital erythropoietic porphyria|HEM4_HUMAN|Hydroxymethylbilane hydrolyase [cyclizing]|Hydroxymethylbilane hydrolyase|OTTHUMP00000020709|OTTHUMP00000020710|UROIIIS|Uroporphyrinogen III cosynthetase|Uroporphyrinogen III synthase (congenital erythropoietic porphyria)|Uroporphyrinogen III synthase|Uroporphyrinogen-III cosynthase|Uroporphyrinogen-III synthase|UROS
- congenital erythropoietic porphyria|HEM4_HUMAN|Hydroxymethylbilane hydrolyase [cyclizing]|Hydroxymethylbilane hydrolyase|OTTHUMP00000020709|OTTHUMP00000020710|UROIIIS|Uroporphyrinogen III cosynthetase|Uroporphyrinogen III synthase (congenital erythropoietic porphyria)|Uroporphyrinogen III synthase|Uroporphyrinogen-III cosynthase|Uroporphyrinogen-III synthase|UROS
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P10746
- P10746
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together