Anti-POLR2E Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: POLR2E
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77794-754
- 77794-753
- Antigen Symbol:
- POLR2E
- POLR2E
- Antigen Name:
- DNA-directed RNA polymerases I, II, and III subunit RPABC1 protein (1-210AA)
- DNA-directed RNA polymerases I, II, and III subunit RPABC1 protein (1-210AA)
- Antigen Synonyms:
- and III subunit ABC1|and III subunit RPABC1|DNA directed RNA polymerase II 23 kda polypeptide|DNA directed RNA polymerase II polypeptide E|DNA directed RNA polymerase II subunit E|DNA directed RNA polymerases I II and III subunit RPABC1|DNA-directed RNA polymerase II 23 kDa polypeptide|DNA-directed RNA polymerase II subunit E|DNA-directed RNA polymerases I|hRPB25|hsRPB5|II|Polymerase (RNA) II (DNA directed) polypeptide E (25kD)|Polymerase (RNA) II (DNA directed) polypeptide E, 25kDa|RNA polymerases I|RNA polymerases I, II, and III subunit ABC1|RPAB1_HUMAN|RPABC1|RPB5|RPB5 homolog|XAP4
- and III subunit ABC1|and III subunit RPABC1|DNA directed RNA polymerase II 23 kda polypeptide|DNA directed RNA polymerase II polypeptide E|DNA directed RNA polymerase II subunit E|DNA directed RNA polymerases I II and III subunit RPABC1|DNA-directed RNA polymerase II 23 kDa polypeptide|DNA-directed RNA polymerase II subunit E|DNA-directed RNA polymerases I|hRPB25|hsRPB5|II|Polymerase (RNA) II (DNA directed) polypeptide E (25kD)|Polymerase (RNA) II (DNA directed) polypeptide E, 25kDa|RNA polymerases I|RNA polymerases I, II, and III subunit ABC1|RPAB1_HUMAN|RPABC1|RPB5|RPB5 homolog|XAP4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P19388
- P19388
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together