Anti-TMEM176B Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: TMEM176B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77802-960
- 77802-961
- Antigen Symbol:
- TMEM176B
- TMEM176B
- Antigen Name:
- Transmembrane protein 176B protein (1-64AA)
- Transmembrane protein 176B protein (1-64AA)
- Antigen Synonyms:
- TMEM176B|LR8|Transmembrane protein 176B|Protein LR8
- TMEM176B|LR8|Transmembrane protein 176B|Protein LR8
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q3YBM2
- Q3YBM2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together