Anti-SACM1L Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SACM1L
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-461
- 77837-460
- Antigen Symbol:
- SACM1L
- SACM1L
- Antigen Name:
- Phosphatidylinositide phosphatase SAC1 protein (301-520AA)
- Phosphatidylinositide phosphatase SAC1 protein (301-520AA)
- Antigen Synonyms:
- Phosphatidylinositide phosphatase SAC1|SAC1|SAC1 suppressor of actin mutations 1-like (yeast)|SAC1_HUMAN|SACM1L|suppressor of actin 1|Suppressor of actin mutations 1-like protein
- Phosphatidylinositide phosphatase SAC1|SAC1|SAC1 suppressor of actin mutations 1-like (yeast)|SAC1_HUMAN|SACM1L|suppressor of actin 1|Suppressor of actin mutations 1-like protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NTJ5
- Q9NTJ5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together