Anti-LNX2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: LNX2
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-337
- 77833-336
- Antigen Symbol:
- LNX2
- LNX2
- Antigen Name:
- Ligand of Numb protein X 2 protein (201-500AA)
- Ligand of Numb protein X 2 protein (201-500AA)
- Antigen Synonyms:
- FLJ12933| FLJ23932| FLJ38000| Ligand of Numb protein X 2| LNX 2| LNX2| LNX2_HUMAN| MGC46315| Numb binding protein 2| Numb-binding protein 2| PDZ domain containing ring finger 1| PDZ domain containing RING finger protein 1| PDZ domain-containing RING finger protein 1| PDZRN 1| PDZRN1
- FLJ12933| FLJ23932| FLJ38000| Ligand of Numb protein X 2| LNX 2| LNX2| LNX2_HUMAN| MGC46315| Numb binding protein 2| Numb-binding protein 2| PDZ domain containing ring finger 1| PDZ domain containing RING finger protein 1| PDZ domain-containing RING finger protein 1| PDZRN 1| PDZRN1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N448
- Q8N448
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together