Anti-SLC18A1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC18A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-783
- 77799-782
- Antigen Symbol:
- SLC18A1
- SLC18A1
- Antigen Name:
- Chromaffin granule amine transporter protein (66-120AA)
- Chromaffin granule amine transporter protein (66-120AA)
- Antigen Synonyms:
- 4832416I10Rik|Adrenal chromaffin granule amine transporter, CGAT|CGAT|Chromaffin granule amine transporter|MGC28683|MGC37299|SLC18A1|Solute carrier family 18 (vesicular monoamine), member 1|Solute carrier family 18 member 1|VAT 1|VAT1|Vesicular amine transporter 1|VMAT 1|VMAT1_HUMAN
- 4832416I10Rik|Adrenal chromaffin granule amine transporter, CGAT|CGAT|Chromaffin granule amine transporter|MGC28683|MGC37299|SLC18A1|Solute carrier family 18 (vesicular monoamine), member 1|Solute carrier family 18 member 1|VAT 1|VAT1|Vesicular amine transporter 1|VMAT 1|VMAT1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P54219
- P54219
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together