Anti-PPP2R2B Rabbit Polyclonal Antibody
Catalog # 77795-115
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B beta isoform
- Antigen Symbol:PPP2R2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q00005
- Antigen Synonyms:2ABB_HUMAN|B55 beta|Beta isoform of regulatory subunit B55 protein phosphatase 2|MGC24888|PP2A B55beta|PP2A PR55B|PP2A subunit B B beta isoform|PP2A subunit B B55 beta isoform|PP2A subunit B isoform B55 beta|PP2A subunit B isoform B55-beta|PP2A subunit B isoform beta|PP2A subunit B isoform PR55 beta|PP2A subunit B isoform PR55-beta|PP2A subunit B isoform R2 beta|PP2A subunit B isoform R2-beta|PP2A subunit B PR55 beta isoform|PP2A subunit B R2 beta isoform|Ppp2r2b|PR2AB beta|PR2AB55 beta|PR2APR55 beta|PR52B|PR55 beta|Protein phosphatase 2 (formerly 2A) regulatory subunit B (PR 52) beta isoform|Protein phosphatase 2 (formerly 2A) regulatory subunit B beta isoform|Protein phosphatase 2 regulatory subunit B|Protein phosphatase 2 regulatory subunit B beta isoform|SCA 12|SCA12|Serine/threonine protein phosphatase 2A 55 kDa regulatory subunit B beta isoform|Serine/threonine protein phosphatase 2A neuronal isoform|Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B beta isoform
- Storage Buffer:PBS, 0.1% Sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: PPP2R2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat