Anti-MAPKAP1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:500 to 1:2000
Type:
Antigen: MAPKAP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-435
- 77834-434
- Antigen Symbol:
- MAPKAP 1;MAPKAP1;MEKK2 interacting protein 1;MGC2745;MIP 1;MIP1;Mitogen activated protein kinase associated protein 1;Mitogen-activated protein kinase 2-associated protein 1;mSIN1;OTTHUMP00000064207;Ras inhibitor MGC2745;SAPK interacting protein 1;SAPK-interacting protein 1;SIN 1;SIN1_HUMAN;SIN1b;SIN1g;Stress activated map kinase interacting protein 1;Stress activated protein kinase interacting 1;Stress-activated map kinase-interacting protein 1;Target of rapamycin complex 2 subunit MAPKAP1;TORC2 subunit MAPKAP1
- MAPKAP 1;MAPKAP1;MEKK2 interacting protein 1;MGC2745;MIP 1;MIP1;Mitogen activated protein kinase associated protein 1;Mitogen-activated protein kinase 2-associated protein 1;mSIN1;OTTHUMP00000064207;Ras inhibitor MGC2745;SAPK interacting protein 1;SAPK-interacting protein 1;SIN 1;SIN1_HUMAN;SIN1b;SIN1g;Stress activated map kinase interacting protein 1;Stress activated protein kinase interacting 1;Stress-activated map kinase-interacting protein 1;Target of rapamycin complex 2 subunit MAPKAP1;TORC2 subunit MAPKAP1
- Antigen Name:
- Target of rapamycin complex 2 subunit MAPKAP1 protein (1-290AA)
- Target of rapamycin complex 2 subunit MAPKAP1 protein (1-290AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BPZ7
- Q9BPZ7
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together