Anti-DCAF7 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: DCAF7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77804-549
- 77804-548
- Antigen Symbol:
- DCAF7
- DCAF7
- Antigen Name:
- DDB1- and CUL4-associated factor 7 protein (1-342AA)
- DDB1- and CUL4-associated factor 7 protein (1-342AA)
- Antigen Synonyms:
- AN11|Dcaf7|DCAF7_HUMAN|DDB1- and CUL4-associated factor 7|HAN11|Human anthocyanin|Petunia|Seven WD repeat protein of the AN11 family 1|SWAN 1|WD repeat domain 68|WD repeat protein An11 homolog|WD repeat-containing protein 68|WD repeat-containing protein An11 homolog|WDR68
- AN11|Dcaf7|DCAF7_HUMAN|DDB1- and CUL4-associated factor 7|HAN11|Human anthocyanin|Petunia|Seven WD repeat protein of the AN11 family 1|SWAN 1|WD repeat domain 68|WD repeat protein An11 homolog|WD repeat-containing protein 68|WD repeat-containing protein An11 homolog|WDR68
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P61962
- P61962
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together