Anti-EPM2AIP1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: EPM2AIP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77831-056
- 77831-057
- Antigen Symbol:
- EPM2AIP1
- EPM2AIP1
- Antigen Name:
- EPM2A-interacting protein 1 protein (251-607AA)
- EPM2A-interacting protein 1 protein (251-607AA)
- Antigen Synonyms:
- EPM2AIP 1| EPM2A (laforin) interacting protein 1| EPM2A (laforin) IP1| EPM2A-interacting protein 1| Epm2aip1| EPMIP_HUMAN| FLJ11207| KIAA0766| Laforin interacting protein 1| Laforin-interacting protein| My007
- EPM2AIP 1| EPM2A (laforin) interacting protein 1| EPM2A (laforin) IP1| EPM2A-interacting protein 1| Epm2aip1| EPMIP_HUMAN| FLJ11207| KIAA0766| Laforin interacting protein 1| Laforin-interacting protein| My007
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7L775
- Q7L775
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together