Anti-TRMT112 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: TRMT112
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-457
- 77841-458
- Antigen Symbol:
- TRMT112
- TRMT112
- Antigen Name:
- tRNA methyltransferase 112 homolog protein (1-125AA)
- tRNA methyltransferase 112 homolog protein (1-125AA)
- Antigen Synonyms:
- TRMT112|AD-001|HSPC152|HSPC170|Multifunctional methyltransferase subunit TRM112-like protein|tRNA methyltransferase 112 homolog
- TRMT112|AD-001|HSPC152|HSPC170|Multifunctional methyltransferase subunit TRM112-like protein|tRNA methyltransferase 112 homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UI30
- Q9UI30
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together