Anti-RAD23B Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RAD23B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77796-563
- 77796-562
- Antigen Symbol:
- RAD23B
- RAD23B
- Antigen Name:
- UV excision repair protein RAD23 homolog B protein (1-250AA)
- UV excision repair protein RAD23 homolog B protein (1-250AA)
- Antigen Synonyms:
- hHR 23b|hHR23B|HR 23B|HR23B|mHR 23B|mHR23B|p58|RAD 23B|RAD23 (S. cerevisiae) homolog B|RAD23 homolog B (S. cerevisiae)|RAD23 homolog B|RAD23 yeast homolog of B|Rad23b|RD23B_HUMAN|UV excision repair protein RAD23 homolog B|XP C repair complementing complex 58 kDa|XP C repair complementing complex 58 kDa protein|XP C repair complementing protein|XP-C repair-complementing complex 58 kDa protein|XPC repair complementing complex 58 kDa|XPC repair complementing complex 58 kDa protein|XPC repair complementing protein
- hHR 23b|hHR23B|HR 23B|HR23B|mHR 23B|mHR23B|p58|RAD 23B|RAD23 (S. cerevisiae) homolog B|RAD23 homolog B (S. cerevisiae)|RAD23 homolog B|RAD23 yeast homolog of B|Rad23b|RD23B_HUMAN|UV excision repair protein RAD23 homolog B|XP C repair complementing complex 58 kDa|XP C repair complementing complex 58 kDa protein|XP C repair complementing protein|XP-C repair-complementing complex 58 kDa protein|XPC repair complementing complex 58 kDa|XPC repair complementing complex 58 kDa protein|XPC repair complementing protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P54727
- P54727
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together