Anti-UGT2B4 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UGT2B4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-978
- 77803-978
- Antigen Symbol:
- UGT2B4
- UGT2B4
- Antigen Name:
- UDP-glucuronosyltransferase 2B4 protein (281-481AA)
- UDP-glucuronosyltransferase 2B4 protein (281-481AA)
- Antigen Synonyms:
- UGT2B4|UGT2B11|UDP-glucuronosyltransferase 2B4|UDPGT 2B4|EC 2.4.1.17|HLUG25|Hyodeoxycholic acid-specific UDPGT|UDPGTh-1
- UGT2B4|UGT2B11|UDP-glucuronosyltransferase 2B4|UDPGT 2B4|EC 2.4.1.17|HLUG25|Hyodeoxycholic acid-specific UDPGT|UDPGTh-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P06133
- P06133
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together