Anti-FKBP8 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FKBP8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-076
- 77823-075
- Antigen Symbol:
- FKBP8
- FKBP8
- Antigen Name:
- Peptidyl-prolyl cis-trans isomerase FKBP8 protein (58-190AA)
- Peptidyl-prolyl cis-trans isomerase FKBP8 protein (58-190AA)
- Antigen Synonyms:
- 38 kDa FK506-binding protein|38 kDa FKBP|38kDa|FK506 binding protein 8|FK506 binding protein 8, 38kDa|FK506 binding protein, 38kDa|FK506-binding protein 8|FKBP 38|FKBP-38|FKBP-8|Fkbp8|FKBP8_HUMAN|FKBPR38|hFKBP38|Peptidyl-prolyl cis-trans isomerase FKBP8|PPIase FKBP8|Rotamase|Sam11
- 38 kDa FK506-binding protein|38 kDa FKBP|38kDa|FK506 binding protein 8|FK506 binding protein 8, 38kDa|FK506 binding protein, 38kDa|FK506-binding protein 8|FKBP 38|FKBP-38|FKBP-8|Fkbp8|FKBP8_HUMAN|FKBPR38|hFKBP38|Peptidyl-prolyl cis-trans isomerase FKBP8|PPIase FKBP8|Rotamase|Sam11
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14318
- Q14318
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together