Anti-PTPA Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:1000 to 1:2000, IF: 1:200 to 1:500
Type:
Antigen: PTPA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-095
- 77824-094
- Antigen Symbol:
- PTPA
- PTPA
- Antigen Name:
- Serine/threonine-protein phosphatase 2A activator protein (1-250AA)
- Serine/threonine-protein phosphatase 2A activator protein (1-250AA)
- Antigen Synonyms:
- subunit B''|KIAA0044|MGC2184|Phosphotyrosyl phosphatase activator|PP2A|PP2A phosphatase activator|PP2A subunit B'|PP2A subunit B' isoform PR53|PP2A subunit B' PR53 isoform|PPP2R4|PR53|PR53 isoform|Protein phosphatase 2, regulatory subunit B-prime|Protein phosphatase 2A activator regulatory subunit 4|Protein phosphatase 2A regulatory subunit B' (PR 53)|PTPA|PTPA_HUMAN|Serine/threonine protein phosphatase 2A regulatory subunit B'|Serine/threonine-protein phosphatase 2A activator|Serine/threonine-protein phosphatase 2A regulatory subunit 4|Serine/threonine-protein phosphatase 2A regulatory subunit B''
- subunit B''|KIAA0044|MGC2184|Phosphotyrosyl phosphatase activator|PP2A|PP2A phosphatase activator|PP2A subunit B'|PP2A subunit B' isoform PR53|PP2A subunit B' PR53 isoform|PPP2R4|PR53|PR53 isoform|Protein phosphatase 2, regulatory subunit B-prime|Protein phosphatase 2A activator regulatory subunit 4|Protein phosphatase 2A regulatory subunit B' (PR 53)|PTPA|PTPA_HUMAN|Serine/threonine protein phosphatase 2A regulatory subunit B'|Serine/threonine-protein phosphatase 2A activator|Serine/threonine-protein phosphatase 2A regulatory subunit 4|Serine/threonine-protein phosphatase 2A regulatory subunit B''
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15257
- Q15257
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together