Order Entry
United States
ContactUsLinkComponent
Anti-BCAM Rabbit Recombinant Polyclonal Antibody
Anti-BCAM Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-BCAM Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77816-377
  • 77816-378
  • Antigen Symbol:
  • BCAM
  • BCAM
  • Antigen Name:
  • Basal cell adhesion molecule protein (447-547AA)
  • Basal cell adhesion molecule protein (447-547AA)
  • Antigen Synonyms:
  • Antigen identified by monoclonal F8|AU|Auberger B antigen|B CAM cell surface glycoprotein|B cell adhesion molecule|B-CAM cell surface glycoprotein|Basal cell adhesion molecule (Lu and Au blood groups)|Basal cell adhesion molecule (Lutheran blood group)|Basal cell adhesion molecule|Basal cell adhesion molecule Lu and Au blood groups|Basal cell adhesion molecule Lutheran blood group|BCAM_HUMAN|CD239|CD239 antigen|F8/G253 antigen|Glycoprotein 95kDa|LU|Lutheran|Lutheran antigen|Lutheran blood group (Auberger b antigen included)|Lutheran blood group Auberger b antigen included|Lutheran blood group glycoprotein|MSK19
  • Antigen identified by monoclonal F8|AU|Auberger B antigen|B CAM cell surface glycoprotein|B cell adhesion molecule|B-CAM cell surface glycoprotein|Basal cell adhesion molecule (Lu and Au blood groups)|Basal cell adhesion molecule (Lutheran blood group)|Basal cell adhesion molecule|Basal cell adhesion molecule Lu and Au blood groups|Basal cell adhesion molecule Lutheran blood group|BCAM_HUMAN|CD239|CD239 antigen|F8/G253 antigen|Glycoprotein 95kDa|LU|Lutheran|Lutheran antigen|Lutheran blood group (Auberger b antigen included)|Lutheran blood group Auberger b antigen included|Lutheran blood group glycoprotein|MSK19
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P50895
  • P50895
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together